Methylenediphosphonic acid 98%

Danger

AC21507-0050 AC21507-0010
103531-820EA 219 USD
103531-820 103531-818
Methylenediphosphonic acid 98%
Methylenediphosphonic acid
Formula: CH₆O₆P₂
Boiling Pt: 82 °C (6 mmHg)
Melting Pt: 200…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00002139
CAS Number: 1984-15-2
EINECS: 217-851-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 197 to 204 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR