Trifluoroacetamidine 85%

Danger

Synonyms: 2,2,2-Trifluoroacetamidine

AC45469-0050 AC45469-0250 AC45469-0010
103531-180EA 43.3 USD
103531-180 103531-182 103531-184
Trifluoroacetamidine 85%
Trifluoroacetamidine
Formula: C₂H₃F₃N₂
MW: 112.05 g/mol
Boiling Pt: 35…37 °C (11 mmHg)
Storage Temperature: Freezer
MDL Number: MFCD00041545
CAS Number: 354-37-0
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to straw to pink Liquid
Infrared spectrum Conforms
HPLC ≥84.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR