N-Methylisopropylamine 98%

Danger

Synonyms: N-Isopropylmethylamine

AC43965-0100 AC43965-0500
103530-706EA 72.4 USD
103530-706 103530-708
N-Methylisopropylamine 98%
N-Methylisopropylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 50…53 °C (760 mmHg)
Density: 0.702 g/cm³
Flash Pt: –32 °C
Storage Temperature: Ambient
MDL Number: MFCD00042859
UN: 2734
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20 °C, 589 nm)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR