Benzyl chloroformate 50% (w/w) 3 M in toluene, AcroSeal™

Danger

Synonyms: CBZ-Cl (Z-Cl)

AC42890-1000
103529-680EA 73.62 USD
103529-680
Benzyl chloroformate 50% (w/w) 3 M in toluene, AcroSeal™
Benzyl chloroformate
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Density: 1.01 g/cm³
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000640
CAS Number: 501-53-1
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear light yellow to light pink Solution
GC 45 to 53 % w/w

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR