cis-2,3-Epoxybutane 97%

Danger

AC30787-0010
103532-252EA 46.1 USD
103532-252
cis-2,3-Epoxybutane 97%
cis-2,3-Epoxybutane
Formula: C₄H₈O
Boiling Pt: 60…61 °C
Melting Pt: 84…83 °C
Density: 0.826 g/cm³
Flash Pt: –21.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005128
CAS Number: 1758-33-4
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR