2-Chloro-3-formylpyridine 98%

Warning

Synonyms: 2-Chloropyridine-3-carboxaldehyde2-Chloronicotinaldehyde2-Chloro-3-pyridinecarboxaldehyde

AC42979-0050 AC42979-0010
103533-256EA 30.9 USD
103533-256 103533-258
2-Chloro-3-formylpyridine 98%
2-Chloro-3-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD01315308
CAS Number: 36404-88-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder or solid
Infrared spectrum Conforms
Melting point 47 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR