Triethyloxonium tetrafluoroborate 1M in methylene chloride, AcroSeal™

Danger

AC42916-1000
103533-208EA 137.31 USD
103533-208
Triethyloxonium tetrafluoroborate 1M in methylene chloride, AcroSeal™
Triethyloxonium tetrafluoroborate
Formula: C₆H₁₅BF₄O
Density: 1.328 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD00044423
CAS Number: 368-39-8
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to yellow to pink to brown Solution (May develop some turbidity or precipitate)
Titration with NaOH 0.90 to 1.10 M
Refractive index 1.4040 to 1.4060 (20 °C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR