Tin (II) trifluoromethanesulfonate 98%

Danger

Synonyms: Tin trifluoromethanesulfonateTin(II) trifluoromethanesulfonateTin triflateTin(II) trifluoromethanesulphonateTin trifluoromethanesulphonate

AC31253-0010 AC31253-0050
103532-330EA 164 USD
103532-330 103532-328
Tin (II) trifluoromethanesulfonate 98%
Tin (II) trifluoromethanesulphonate
Formula: C₂F₆O₆S₂Sn
MW: 416.85 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00191251
CAS Number: 62086-04-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR