4-Chlorophenethyl bromide 95%

Warning

Synonyms: 1-(2-Bromoethyl)-4-chlorobenzene

AC44018-0050
103533-676EA 106 USD
103533-676
4-Chlorophenethyl bromide 95%
4-Chlorophenethyl bromide
Formula: C₈H₈BrCl
MW: 219.51 g/mol
Boiling Pt: 85 °C (1Torr)
Density: 1.508 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00634271
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR