2-Chlorophenethyl bromide 95%

Danger

Synonyms: 2-Chlorophenethyl bromide2-(2-Chlorophenyl)ethyl bromide1-(2-Bromoethyl)-2-chlorobenzene2-Chlorophenethyl Bromide

AC44014-0050 AC44014-0010
103533-670EA 130 USD
103533-670 103533-668
2-Chlorophenethyl bromide 95%
2-Chlorophenethyl bromide
Formula: C₈H₈BrCl
MW: 219.51 g/mol
Boiling Pt: 110…112 °C (10Torr)
Density: 1.4569 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04117464
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR