2-Bromo-3,3,3-trifluoropropene 97%

Warning

Synonyms: 2-Bromo-3,3,3-trifluoro-1-propene

AC43446-0010 AC43446-0050
103533-426EA 43.9 USD
103533-426 103533-428
2-Bromo-3,3,3-trifluoropropene 97%
2-Bromo-3,3,3-trifluoropropene
Formula: C₃H₂BrF₃
MW: 174.95 g/mol
Boiling Pt: 34 °C (760 mmHg)
Density: 1.65 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00077469
CAS Number: 1514-82-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3520 to 1.3570 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR