2-Methoxyphenethylamine 98%

Danger

AC18217-0050 AC18217-1000 AC18217-0250
103531-700EA 97.1 USD
103531-700 103531-702 103531-698
2-Methoxyphenethylamine 98%
2-Methoxyphenethylamine
Formula: C₉H₁₃NO
Boiling Pt: 236…237 °C
Density: 1.03 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008186
CAS Number: 2045-79-6
EINECS: 218-066-6
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR