2-Amino-4-phenylthiazole 97%
|
Danger
|
Synonyms:
4-Phenyl-1,3-thiazol-2-amine
, 4-Phenylthiazol-2-amine
AC43249-0010
AC43249-0050
103533-328EA
186
USD
103533-328
103533-326
2-Amino-4-phenylthiazole 97%
2-Amino-4-phenylthiazole
Formula:
C₉H₈N₂S MW: 176.24 g/mol Melting Pt: 150…152.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039680 CAS Number: 2010-06-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 149 to 153 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...