2,5-Dihydrofuran 98%

Danger

AC24478-0250 AC24478-1000
103531-974EA 59.7 USD
103531-974 103531-976
2,5-Dihydrofuran 98%
2,5-Dihydrofuran
Formula: C₄H₆O
Boiling Pt: 66…67 °C (760mbar)
Density: 0.92 g/cm³
Flash Pt: –016 °C
Storage Temperature: Ambient
MDL Number: MFCD00003219
CAS Number: 1708-29-8
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR