3-Fluorobenzenesulfonamide 97%

Danger

AC43784-0010
103530-618EA 108 USD
103530-618
3-Fluorobenzenesulfonamide 97%
3-Fluorobenzenesulphonamide
Formula: C₆H₆FNO₂S
MW: 175.18 g/mol
Melting Pt: 131.1…132.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00042284
CAS Number: 1524-40-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 128.0 to 134.0 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR