3-Amino-5-chloropyridine 95%

Danger

Synonyms: 5-Chloropyridin-3-amine

AC44890-0050 AC44890-0010 AC44890-2500
103531-106EA 480 USD
103531-106 103531-108 103531-104
3-Amino-5-chloropyridine 95%
3-Amino-5-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 51…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03701386
CAS Number: 22353-34-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Cream to beige to light brown Powder
Infrared spectrum Conforms
Melting point 71 to 75 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR