Phenylacetyl chloride 98%

Danger

AC13030-1000 AC13030-5000
103530-892EA 42.3 USD
103530-892 103530-894
Phenylacetyl chloride 98%
Phenylacetyl chloride
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 94…95 °C (12 mmHg)
Melting Pt: 91 °C
Density: 1.16 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00000729
CAS Number: 103-80-0
EINECS: 203-146-5
UN: 2577
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow to pink-red Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5330 (20 °C, 589 nm)
GC ≥97.5 %
Titration RCOCl ≥97.5 %
Titration RCOOH ≤2 %
Free acid (HCl) Traces
Color scale ≤20 APHA
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR