2-Bromonicotinic acid 97%
|
Warning
|
Synonyms:
2-Bromo-3-pyridinecarboxylic acid
, 2-Bromopyridine-3-carboxylic acid
AC42977-0050
AC42977-0010
103533-250EA
127
USD
103533-250
103533-248
2-Bromonicotinic acid 97%
2-Bromonicotinic acid
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Melting Pt: 200…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646068 CAS Number: 35905-85-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to tan Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...