N,N'-Diethylethylenediamine 95%

Danger

AC15552-0250 AC15552-0050
103531-426EA 50.2 USD
103531-426 103531-428
N,N'-Diethylethylenediamine 95%
N,N'-Diethylethylenediamine
Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 152…154 °C (760 mmHg)
Melting Pt: –11 °C
Density: 0.811 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009033
CAS Number: 111-74-0
UN: 2685
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR