3-Bromoisonicotinonitrile 97%

Danger

AC45227-0050 AC45227-0010
103531-160EA 367.7 USD
103531-160 103531-162
3-Bromoisonicotinonitrile 97%
3-Bromoisonicotinonitrile
Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00234143
CAS Number: 13958-98-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Powder or solid
Infrared spectrum Conforms
Melting point 91 to 97 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR