4-Fluorobenzyl mercaptan 96%

Warning

Synonyms: 4-Fluorobenzylmercaptanp-Fluorotoluene-α-thiol

AC44390-0010
103533-842EA 51.3 USD
103533-842
4-Fluorobenzyl mercaptan 96%
4-Fluorobenzyl mercaptan
Formula: C₇H₇FS
MW: 142.2 g/mol
Boiling Pt: 72…74 °C (15 mmHg)
Density: 1.157 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00040724
CAS Number: 15894-04-9
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR