Cypermethrin 98%

Danger

AC45631-0250 AC45631-0050 AC45631-1000
103534-284EA 167 USD
103534-284 103534-288 103534-286
Cypermethrin 98%
Cypermethrin
Formula: C₂₂H₁₉Cl₂NO₃
MW: 416.3 g/mol
Density: 1.12 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00055328
CAS Number: 52315-07-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
HPLC ≥97.5 % (Mixture of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR