2,4-Dimethoxyphenyl isothiocyanate 95%
|
Danger
|
Synonyms:
1,3-Dimethoxy-4-isothiocyanatobenzene
, 2,4-Dimethoxyphenylisothiocyanate
AC44178-0050
AC44178-0010
103533-716EA
56.6
USD
103533-716
103533-714
2,4-Dimethoxyphenyl isothiocyanate 95%
2,4-Dimethoxyphenyl isothiocyanate
Formula:
C₉H₉NO₂S MW: 195.24 g/mol Boiling Pt: 166…167 °C (6 mmHg) Melting Pt: 54…55 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041058 CAS Number: 33904-03-9 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | Light yellow Powder or low melting solid |
Infrared spectrum | Conforms |
Melting point | 52 to 56 °C |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...