3,5-Difluorobenzenesulfonyl chloride 97%

Danger

Synonyms: 3,5-Difluorobenzene-1-sulfonyl chloride3,5-Difluorobenzene-1-sulphonyl chloride

AC43100-0010 AC43100-0050
103529-736EA 43.8 USD
103529-736 103529-738
3,5-Difluorobenzenesulfonyl chloride 97%
3,5-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Melting Pt: 55.9…58.2 °C
Storage Temperature: Ambient
MDL Number: MFCD02091378
CAS Number: 210532-25-5
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Orange to tan Crystalline powder
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR