2-Fluorobenzenesulfonyl chloride 97%

Danger

Synonyms: 2-Fluorobenzene-1-sulfonyl chloride2-Fluorobenzene-1-sulphonyl chloride

AC37578-0010 AC37578-0050
103529-614EA 25 USD
103529-614 103529-616
2-Fluorobenzenesulfonyl chloride 97%
2-Fluorobenzenesulphonyl chloride
Formula: C₆H₄ClFO₂S
MW: 194.61 g/mol
Boiling Pt: 140…142 °C (12 mmHg)
Melting Pt: 26 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042285
CAS Number: 2905-21-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5390 (20 °C, 589 nm)
Titration Argentometric ≥96.0 % (ex Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR