2-Fluoroethanol 95%

Danger

AC17083-0050 AC17083-0250
103531-658EA 68.06 USD
103531-658 103531-660
2-Fluoroethanol 95%
2-Fluoroethanol
Formula: C₂H₅FO
MW: 64.06 g/mol
Boiling Pt: 100…104 °C (760 mmHg)
Melting Pt: –26.5 °C
Density: 1.108 g/cm³
Flash Pt: 34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002828
CAS Number: 371-62-0
UN: 3383
ADR: 6.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR