p-Toluenesulfonic acid monohydrate ACS

Danger

Synonyms: 4-Methylbenzenesulphonic acid hydrate4-Methylbenzenesulfonic acid hydrateToluene-4-sulphonic acid monohydrateToluene-4-sulfonic acid monohydrate

AC42121-0250
103533-128EA 22.5 USD
103533-128
p-Toluenesulfonic acid monohydrate ACS
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Melting Pt: 100…108 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
UN: 2585
ADR: 8,III

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Water 9.5 to 11.5 %
Sulfated ash ≤0.1 %
Titration with NaOH ≥98.5 %
Clarity of solution (c=20, water) Passes test
Sulfate (SO4) ≤0.3 %
Iron (Fe) ≤0.01 %
Sodium (Na) ≤0.002 %
Heavy metals (as Pb) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR