(±)-3-Quinuclidinol 99%

Danger

AC13224-0050 AC13224-0250
103530-942EA 112 USD
103530-942 103530-940
(±)-3-Quinuclidinol 99%
(±)-3-Quinuclidinol
Formula: C₇H₁₃NO
MW: 127.19 g/mol
Melting Pt: 217…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00151326
CAS Number: 1619-34-7
EINECS: 216-578-4
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 217 to 224 °C
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR