Propionyl chloride 99.5% phosgene-free

Danger

AC45897-5000 AC45897-0250
103531-228EA 52.2 USD
103531-228 103529-964
Propionyl chloride 99.5% phosgene-free
Propionyl chloride
Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –94 °C
Density: 1.059 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
UN: 1815
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4050 (20 °C, 589 nm)
GC ≥99.4 %
Color scale ≤20 APHA
Titration RCOCl ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR