5-Amino-2-methylpyridine 97%

Danger

Synonyms: 3­Amino­6­methylpyridine6-Methylpyridin-3-amine6-Methyl-3-pyridinamine5-Amino-2-picoline3­Amino­6­picoline

AC43668-0010
103529-836EA 188.22 USD
103529-836
5-Amino-2-methylpyridine 97%
5-Amino-2-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 130 °C (1Torr)
Melting Pt: 93.6…94.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00833389
CAS Number: 3430-14-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to orange to brown Powder
Infrared spectrum Conforms
Melting point 93 to 99 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR