2,3-Difluorobenzoyl chloride 98%

Danger

Synonyms: 2,3-Difluorobenzoylchloride

AC37582-0010
103529-618EA 24.7 USD
103529-618
2,3-Difluorobenzoyl chloride 98%
2,3-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 85…87 °C (14 mmHg)
Density: 1.423 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00042499
CAS Number: 18355-73-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5140 to 1.5160 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR