1-Bromo-2,3,5-trifluorobenzene 98%

Warning

Synonyms: 2,3,5-Trifluorobromobenzene

AC44384-0050 AC44384-0010
103533-834EA 29.8 USD
103533-834 103533-836
1-Bromo-2,3,5-trifluorobenzene 98%
1-Bromo-2,3,5-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 143 °C (760 mmHg)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00012232
CAS Number: 133739-70-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR