4-Bromo-m-toluidine 97%

Warning

Synonyms: 5-Amino-2-bromotoluene4-Bromo-3-methylaniline3-Amino-6-bromotoluene

AC16750-2500 AC16750-0100
103531-602EA 344 USD
103531-602 103531-598
4-Bromo-m-toluidine 97%
4-Bromo-m-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 240 °C (760 mmHg)
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007828
CAS Number: 6933-10-4
EINECS: 230-056-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR