1,5-Dihydroxynaphthalene 97%

Warning

Synonyms: Naphthalene-1,5-diol1,5-Naphthalenediol

AC11500-1000 AC11500-5000
103530-556EA 164 USD
103530-556 103530-554
1,5-Dihydroxynaphthalene 97%
1,5-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 259…261 °C (dec.)
Flash Pt: 252 °C
Storage Temperature: Ambient
MDL Number: MFCD00003980
CAS Number: 83-56-7
EINECS: 201-487-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Grey-brown to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR