Indole-4-carbonitrile 97%

Danger

Synonyms: 1H-Indole-4-carbonitrile4-Cyano-1H-indole4-Cyanoindole

AC44249-0010
103533-772EA 42.1 USD
103533-772
Indole-4-carbonitrile 97%
Indole-4-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 120…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00152045
CAS Number: 16136-52-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
Melting point 118 to 122 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR