Mercury(I) chloride ACS

Danger

Synonyms: Mercurous chlorideDimercury dichlorideCalomel

AC44759-5000 AC44759-1000
103533-938EA 91.7 USD
103533-938 103533-940
Mercury(I) chloride ACS
Mercury (I) chloride
Formula: Cl₂Hg₂
MW: 472.09 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 400 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011043
CAS Number: 10112-91-1
UN: 2025
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Titration Iodometric ≥99.5 %
Sulfate (SO4) ≤0.01 %
Mercuric Chloride ≤0.01 % (HgCl2)
Residue after reduction ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR