3-Bromobenzylamine 95%

Danger

Synonyms: (3-Bromophenyl)methanamine

AC42896-0250 AC42896-0010 AC42896-0050
103529-688EA 29.2 USD
103529-688 103529-690 103529-692
3-Bromobenzylamine 95%
3-Bromobenzylamine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01026119
CAS Number: 10269-01-9
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5850 to 1.5900 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR