2-Hydroxythiophenol 98+%

Danger

Synonyms: o-Hydroxythiophenol2-Mercaptophenol1-Hydroxy-2-mercaptobenzene2-Hydroxyphenyl mercaptan2-Hydroxybenzene-1-thiol2-Hydroxybenzenethiol

AC38560-0010 AC38560-0050
103532-872EA 50.2 USD
103532-872 103532-874
2-Hydroxythiophenol 98+%
2-Hydroxythiophenol
Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 40 °C (300 mmHg)
Melting Pt: 20 °C
Density: 1.22 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00040447
CAS Number: 1121-24-0
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear yellow to light green Liquid
Infrared spectrum Conforms
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR