Cadmium(II) hydroxide (≥75% Cd), pure
|
Danger
|
AC45388-0010
AC45388-2500
103534-184EA
245.5
USD
103534-184
103534-186
Cadmium(II) hydroxide (≥75% Cd), pure
Cadmium (II) hydroxide
Formula:
H₂CdO₂ MW: 146.42 g/mol Melting Pt: 232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015995 CAS Number: 21041-95-2 UN: 2570 ADR: 6.1,III |
Specification Test Results
Appearance | White Powder |
Titration excess HCl | ≥70 % (Cd) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...