Bismuth(III) nitrate pentahydrate 98%, pure

Danger

Synonyms: Bismuth trinitrate pentahydrate

AC45411-0010 AC45411-0050
103534-200EA 113 USD
103534-200 103534-202
Bismuth(III) nitrate pentahydrate 98%, pure
Bismuth (III) nitrate pentahydrate
Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00149157
CAS Number: 10035-06-0
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR