Bismuth(III) nitrate pentahydrate ACS

Danger

Synonyms: Bismuth trinitrate pentahydrate

AC44756-5000 AC44756-1000
103533-932EA 124 USD
103533-932 103533-930
Bismuth(III) nitrate pentahydrate ACS
Bismuth (III) nitrate pentahydrate
Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00149157
CAS Number: 10035-06-0
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White or transparent Crystalline powder or crystals and/or chunks
Titration Complexometric ≥98.0 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Arsenic (As) ≤0.001 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.002 %
Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.002 %
Potassium (K) ≤0.01 %
Silver (Ag) ≤0.001 %
Sodium (Na) ≤0.02 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR