Sodium cyanamidate ≥98%

Danger

Synonyms: Sodium hydrogencyanamide

AC43805-0050 AC43805-0250
103533-560EA 124.13 USD
103533-560 103533-558
Sodium cyanamidate ≥98%
Sodium cyanamidate
Formula: CHN₂Na
MW: 64.02 g/mol
Melting Pt: >360 °C
Storage Temperature: Ambient
MDL Number: MFCD00066500
CAS Number: 17292-62-5
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR