Guanidinium thiocyanate 99+% for molecular biology

Danger

Synonyms: Guanidinium sulfocyanideGuanidine ThiocyanateGuanidinium sulphocyanideGTCGuanidinium rhodanide

AC43394-1000
103530-396EA 87.7 USD
103530-396
Guanidinium thiocyanate 99+% for molecular biology
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 118 to 122 °C
Titration Argentometric 99.0 to 101.0 %
Loss on drying ≤0.5 %
Sulfated ash ≤0.05 %
Solubility (3 M in water) Clear colorless
UV at 280 nm A: ≤0.4
UV at 300 nm A: ≤0.05
UV (3 M in water)
pH 5.0 to 7.0 (1 M in water)
Dnase,Rnase,Protease activity None detected
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Zinc (Zn) ≤2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR