Cadmium(II) carbonate, pure

Danger

AC21232-2500 AC21232-0010
103531-798EA 155 USD
103531-798 103531-800
Cadmium(II) carbonate, pure
Cadmium (II) carbonate
Formula: CCdO₃
MW: 172.42 g/mol
Melting Pt: 343…412 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010918
CAS Number: 513-78-0
EINECS: 208-168-9
UN: 2570
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to off-white Powder
Assay 60.0 to 63.0 % (Cd)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR