3-Chlorothiophene 96%

Danger

AC25880-0050 AC25880-0250
103532-008EA 177.7 USD
103532-008 103532-006
3-Chlorothiophene 96%
3-Chlorothiophene
Formula: C₄H₃ClS
MW: 118.59 g/mol
Boiling Pt: 137…139 °C (760 mmHg)
Density: 1.276 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00043887
CAS Number: 17249-80-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR