2,3-Dichlorothiophene 97%

Danger

AC25879-0010 AC25879-0050
103532-004EA 175.1 USD
103532-004 103532-002
2,3-Dichlorothiophene 97%
2,3-Dichlorothiophene
Formula: C₄H₂Cl₂S
MW: 153.03 g/mol
Boiling Pt: 172…174 °C (760 mmHg)
Melting Pt: –37 °C
Density: 1.47 g/cm³
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043890
CAS Number: 17249-79-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR