2-Chloro-3-methyl-5-nitropyridine 98%

Warning

Synonyms: 2-Chloro-5-nitro-3-picoline

AC36093-0010 AC36093-0050
103532-580EA 164.8 USD
103532-580 103532-578
2-Chloro-3-methyl-5-nitropyridine 98%
2-Chloro-3-methyl-5-nitropyridine
Formula: C₆H₅ClN₂O₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00173686
CAS Number: 22280-56-4
EINECS: 244-889-5
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Cream to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 45 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR