Antimony(V) chloride 1M in methylene chloride, AcroSeal™

Danger

Synonyms: Antimony pentachloride

AC43452-8000 AC43452-1000
103533-432EA 100.28 USD
103533-432 103533-434
Antimony(V) chloride 1M in methylene chloride, AcroSeal™
Antimony (V) chloride
Formula: Cl₅Sb
MW: 299.02 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.442 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011213
CAS Number: 7647-18-9
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to dark yellow Solution
Titration Argentometric 0.95 to 1.10 M
Refractive index 1.5310 to 1.5350 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR