Sodium chlorate 99% ACS

Danger

AC44641-5000
103531-016EA 107 USD
103531-016
Sodium chlorate 99% ACS
Sodium chlorate
Formula: ClNaO₃
MW: 106.44 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 248…261 °C
Density: 2.49 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
UN: 1495
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration ≥99.0 %
Chloride (Cl) ≤0.005 %
Bromate (BrO3) ≤0.015 %
Sulfate (SO4) ≤0.001 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.002 %
Potassium (K) ≤0.01 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR