4-tert-Butylcyclohexylamine cis- and trans-mixture 97%

Danger

Synonyms: 4-(tert-Butyl)cyclohexanamine

AC45560-0050 AC45560-0250
103534-250EA 169 USD
103534-250 103534-248
4-tert-Butylcyclohexylamine cis- and trans-mixture 97%
4-tert-Butylcyclohexylamine
Formula: C₁₀H₂₁N
MW: 155.28 g/mol
Density: 0.87 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00013763
CAS Number: 5400-88-4
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4660 (20 °C, 589 nm)
GC ≥96.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR